C12H316CF2BC Chemcia Scientific, LLC

2-Amino-3-methoxy-4-methyl-benzoic acid methyl ester

  • MDL number MFCD17676541
  • Cas Number : 5544-24-1
  • Molecular Weight : 195.22001

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7804

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7804
Availability details for
Quantity
  • Availability
  • Add to Cart