C12H316CF2BE Chemcia Scientific, LLC

4-Fluoro-5-methyl-2-nitro-benzoic acid

  • MDL number MFCD28797613
  • Cas Number : 1437780-11-4
  • Molecular Weight : 199.13972

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7817

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7817
Availability details for
Quantity
  • Availability
  • Add to Cart