C12H316CF2C5 Chemcia Scientific, LLC

6-Bromo-5-chloro-4H-benzo[1,4]oxazin-3-one

  • MDL number MFCD28402357
  • Cas Number : 1154740-66-5
  • Molecular Weight : 262.49155

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Pink Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7855

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7855
Availability details for
Quantity
  • Availability
  • Add to Cart