C12H316CF2C6 Chemcia Scientific, LLC

6-Bromo-5-chloro-3,4-dihydro-2H-benzo[1,4]oxazine

  • MDL number MFCD24387197
  • Cas Number : 1154740-67-6
  • Molecular Weight : 248.50809

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7856

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7856
Availability details for
Quantity
  • Availability
  • Add to Cart