C12H316CF2C7 Chemcia Scientific, LLC

5-Chloro-6-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-3,4-dihydro-2H-benzo[1,4]oxazine

  • MDL number MFCD22627857
  • Cas Number : 1154740-68-7
  • Molecular Weight : 295.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-benzo[b][1,4]oxazine
Inchi Code 1S/C14H19BClNO3/c1-13(2)14(3,4)20-15(19-13)9-5-6-10-12(11(9)16)17-7-8-18-10/h5-6,17H,7-8H2,1-4H3
Inchi Key GWKYJFILHVNGEL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7857

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7857
Availability details for
Quantity
  • Availability
  • Add to Cart