C12H316CF2CE Chemcia Scientific, LLC

4-Chloro-benzofuran-2-carboxylic acid ethyl ester

  • MDL number MFCD22574024
  • Cas Number : 1383842-58-7
  • Molecular Weight : 224.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-chloro-1-benzofuran-2-carboxylate
Inchi Code 1S/C11H9ClO3/c1-2-14-11(13)10-6-7-8(12)4-3-5-9(7)15-10/h3-6H,2H2,1H3
Inchi Key SXBCZGIYEOLDCM-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7874

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7874
Availability details for
Quantity
  • Availability
  • Add to Cart