C12H316CF2D2 Chemcia Scientific, LLC

1-Bromo-5-chloro-4-fluoro-2-iodo-benzene

  • MDL number MFCD12026232
  • Cas Number : 1067882-65-8
  • Molecular Weight : 335.34264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7917

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7917
Availability details for
Quantity
  • Availability
  • Add to Cart