C12H316CF2D8 Chemcia Scientific, LLC

2-Bromomethyl-6-methyl-benzonitrile

  • MDL number MFCD18394357
  • Cas Number : 1261573-68-5
  • Molecular Weight : 210.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(bromomethyl)-6-methylbenzonitrile
Inchi Code 1S/C9H8BrN/c1-7-3-2-4-8(5-10)9(7)6-11/h2-4H,5H2,1H3
Inchi Key AXJWJJNYAVDONP-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7944

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7944
Availability details for
Quantity
  • Availability
  • Add to Cart