C12H316CF2EB Chemcia Scientific, LLC

2-Bromo-4-fluoro-5-methoxy-phenylamine

  • MDL number MFCD09800756
  • Cas Number : 420786-92-1
  • Molecular Weight : 220.04234

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8024

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8024
Availability details for
Quantity
  • Availability
  • Add to Cart