C12H316CF2F0 Chemcia Scientific, LLC

(2-Bromo-pyridin-3-yl)-hydrazine

  • MDL number MFCD26403440
  • Cas Number : 1293407-97-2
  • Molecular Weight : 188.02767

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8030

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8030
Availability details for
Quantity
  • Availability
  • Add to Cart