C12H316CF2F5 Chemcia Scientific, LLC

4,5-Difluoro-2-hydroxy-benzonitrile

  • MDL number MFCD0827528
  • Cas Number : 186590-36-3
  • Molecular Weight : 155.10486

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8046

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8046
Availability details for
Quantity
  • Availability
  • Add to Cart