C12H316CF309 Chemcia Scientific, LLC

1-Bromo-4,5-difluoro-2-nitro-benzene

  • MDL number MFCD06808551
  • Cas Number : 321-17-5
  • Molecular Weight : 237.98914

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8089

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8089
Availability details for
Quantity
  • Availability
  • Add to Cart