C12H316CF30B Chemcia Scientific, LLC

4,5-Difluoro-2-nitro-phenol

  • MDL number MFCD03094187
  • Cas Number : 55346-97-9
  • Molecular Weight : 175.09251

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8094

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8094
Availability details for
Quantity
  • Availability
  • Add to Cart