C12H316CF320 Chemcia Scientific, LLC

5-Hydroxy-4-methoxy-2-nitro-benzaldehyde

  • MDL number MFCD00228444
  • Cas Number : 58749-47-6
  • Molecular Weight : 197.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-hydroxy-4-methoxy-2-nitrobenzaldehyde
Inchi Code 1S/C8H7NO5/c1-14-8-3-6(9(12)13)5(4-10)2-7(8)11/h2-4,11H,1H3
Inchi Key PTULGYAZUWPSQM-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8121

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8121
Availability details for
Quantity
  • Availability
  • Add to Cart