C12H316CF321 Chemcia Scientific, LLC

2-Bromo-4-fluoro-5-methoxy-benzoic acid methyl ester

  • Cas Number : 1007455-22-2
  • Molecular Weight : 263.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-bromo-4-fluoro-5-methoxybenzoate
Inchi Code 1S/C9H8BrFO3/c1-13-8-3-5(9(12)14-2)6(10)4-7(8)11/h3-4H,1-2H3
Inchi Key KDJBAOOGTMLSCK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8124

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8124
Availability details for
Quantity
  • Availability
  • Add to Cart