C12H316CF330 Chemcia Scientific, LLC

(3-Methoxy-2-methyl-phenyl)-carbamic acid tert-butyl ester

  • Cas Number : 164082-77-3
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-methoxy-2-methylphenylcarbamate
Inchi Code 1S/C13H19NO3/c1-9-10(7-6-8-11(9)16-5)14-12(15)17-13(2,3)4/h6-8H,1-5H3,(H,14,15)
Inchi Key RZZFTOJEHIRCRK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8171

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8171
Availability details for
Quantity
  • Availability
  • Add to Cart