C12H316CF332 Chemcia Scientific, LLC

6-Fluoro-4-methyl-pyridine-3-sulfonyl chloride

  • MDL number MFCD30529914
  • Molecular Weight : 209.62765

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8195

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8195
Availability details for
Quantity
  • Availability
  • Add to Cart