C12H316CF334 Chemcia Scientific, LLC

4-Methyl-pyridine-2,5-diamine

  • Cas Number : 6909-93-9
  • Molecular Weight : 123.15873

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Reddish Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8199

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8199
Availability details for
Quantity
  • Availability
  • Add to Cart