C12H316CF33A Chemcia Scientific, LLC

2-Trifluoromethyl-1H-benzoimidazol-5-ol

  • MDL number MFCD01658719
  • Molecular Weight : 202.13705

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8225

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8225
Availability details for
Quantity
  • Availability
  • Add to Cart