C12H316CF33B Chemcia Scientific, LLC

2-Trifluoromethyl-3H-benzoimidazol-5-ol

  • MDL number MFCD28526364
  • Cas Number : 89426-90-4
  • Molecular Weight : 202.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(trifluoromethyl)-1H-benzo[d]imidazol-6-ol
Inchi Code 1S/C8H5F3N2O/c9-8(10,11)7-12-5-2-1-4(14)3-6(5)13-7/h1-3,14H,(H,12,13)
Inchi Key XKWNYYWVRVWOEN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8226

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8226
Availability details for
Quantity
  • Availability
  • Add to Cart