C12H316CF33C Chemcia Scientific, LLC

1-Fluoro-3,5-dimethyl-benzene

  • MDL number MFCD00052366
  • Cas Number : 461-97-2
  • Molecular Weight : 124.15933

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Colorless Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8328

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8328
Availability details for
Quantity
  • Availability
  • Add to Cart