C12H316CF360 Chemcia Scientific, LLC

3-Nitro-pyrrolo[3,2-b]pyridine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD30529927
  • Molecular Weight : 263.25511

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8262

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8262
Availability details for
Quantity
  • Availability
  • Add to Cart