C12H316CF363 Chemcia Scientific, LLC

3-Benzyloxy-5-bromo-2-chloro-pyrazine

  • MDL number MFCD30529933
  • Cas Number : 1410164-69-0
  • Molecular Weight : 299.55621

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8280

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8280
Availability details for
Quantity
  • Availability
  • Add to Cart