C12H316CF367 Chemcia Scientific, LLC

2-Bromo-5,6,7,8-tetrahydro-[1,2,4]triazolo[1,5-a]pyrazine

  • MDL number MFCD18250478
  • Cas Number : 1260663-45-3
  • Molecular Weight : 203.04234

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8291

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8291
Availability details for
Quantity
  • Availability
  • Add to Cart