C12H316CF369 Chemcia Scientific, LLC

(2-Chloro-5-nitro-phenyl)-dimethyl-amine

  • Cas Number : 17815-97-3
  • Molecular Weight : 200.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-N,N-dimethyl-5-nitroaniline
Inchi Code 1S/C8H9ClN2O2/c1-10(2)8-5-6(11(12)13)3-4-7(8)9/h3-5H,1-2H3
Inchi Key NEDWDFYUMIRLQF-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Red Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8295

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8295
Availability details for
Quantity
  • Availability
  • Add to Cart