C12H316CF36D Chemcia Scientific, LLC

1,4-Dichloro-2-methyl-5-nitro-benzene

  • MDL number MFCD00033868
  • Cas Number : 7149-76-0
  • Molecular Weight : 206.0294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8307

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8307
Availability details for
Quantity
  • Availability
  • Add to Cart