C12H316CF36E Chemcia Scientific, LLC

1-(2-Bromo-4-nitro-phenyl)-ethanone

  • MDL number MFCD00152297
  • Cas Number : 90004-93-6
  • Molecular Weight : 244.04592

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8312

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8312
Availability details for
Quantity
  • Availability
  • Add to Cart