C12H316CF370 Chemcia Scientific, LLC

(4-Amino-2-bromo-phenyl)-methanol

  • MDL number MFCD12152196
  • Cas Number : 1179067-35-6
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-amino-2-bromophenyl)methanol
Inchi Code 1S/C7H8BrNO/c8-7-3-6(9)2-1-5(7)4-10/h1-3,10H,4,9H2
Inchi Key CGMBOCGFJOOYSQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8314

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8314
Availability details for
Quantity
  • Availability
  • Add to Cart