C12H316CF372 Chemcia Scientific, LLC

4,5-Diamino-2-chloro-benzonitrile

  • MDL number MFCD20719621
  • Cas Number : 497147-90-7
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,5-diamino-2-chlorobenzonitrile
Inchi Code 1S/C7H6ClN3/c8-5-2-7(11)6(10)1-4(5)3-9/h1-2H,10-11H2
Inchi Key UWSZKJXTQGXNCK-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Red Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8326

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8326
Availability details for
Quantity
  • Availability
  • Add to Cart