C12H316CF38A Chemcia Scientific, LLC

1-Bromo-4-methoxy-5-nitro-2-trifluoromethyl-benzene

  • Molecular Weight : 300.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-4-methoxy-5-nitro-2-(trifluoromethyl)benzene
Inchi Code 1S/C8H5BrF3NO3/c1-16-7-2-4(8(10,11)12)5(9)3-6(7)13(14)15/h2-3H,1H3
Inchi Key SVRSYJPCHJIUBR-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8403

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8403
Availability details for
Quantity
  • Availability
  • Add to Cart