C12H316CF3A1 Chemcia Scientific, LLC

2-Bromo-4-methyl-5-nitro-benzonitrile

  • MDL number MFCD28745272
  • Cas Number : 1202858-65-8
  • Molecular Weight : 241.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-4-methyl-5-nitrobenzonitrile
Inchi Code 1S/C8H5BrN2O2/c1-5-2-7(9)6(4-10)3-8(5)11(12)13/h2-3H,1H3
Inchi Key SQUUGFCETPITLP-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8466

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8466
Availability details for
Quantity
  • Availability
  • Add to Cart