C12H316CF3A7 Chemcia Scientific, LLC

1-Bromo-2-fluoro-3-iodo-4-methoxy-benzene

  • MDL number MFCD30723577
  • Cas Number : 1628450-73-6
  • Molecular Weight : 330.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-2-fluoro-3-iodo-4-methoxybenzene
Inchi Code 1S/C7H5BrFIO/c1-11-5-3-2-4(8)6(9)7(5)10/h2-3H,1H3
Inchi Key ONYIDRWMDNFQOP-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8527

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8527
Availability details for
Quantity
  • Availability
  • Add to Cart