C12H316CF3AC Chemcia Scientific, LLC

3-Amino-4-bromo-thieno[2,3-c]pyridine-2-carboxylic acid methyl ester

  • MDL number MFCD23159537
  • Cas Number : 1572518-01-4
  • Molecular Weight : 287.13634

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8592

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8592
Availability details for
Quantity
  • Availability
  • Add to Cart