C12H316CF3AD Chemcia Scientific, LLC

3-Bromo-6-methoxy-imidazo[1,2-b]pyridazine

  • MDL number MFCD14706290
  • Cas Number : 453548-63-5
  • Molecular Weight : 228.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromoimidazo[1,2-b]pyridazin-6-yl methyl ether
Inchi Code 1S/C7H6BrN3O/c1-12-7-3-2-6-9-4-5(8)11(6)10-7/h2-4H,1H3
Inchi Key MSEJHRBBLYGKTI-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8599

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8599
Availability details for
Quantity
  • Availability
  • Add to Cart