C12H316CF3AE Chemcia Scientific, LLC

2-Formyl-5-methyl-benzoic acid

  • MDL number MFCD20655933
  • Cas Number : 150867-03-1
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-formyl-5-methylbenzoic acid
Inchi Code 1S/C9H8O3/c1-6-2-3-7(5-10)8(4-6)9(11)12/h2-5H,1H3,(H,11,12)
Inchi Key PGJBLPDZCOFENP-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8629

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8629
Availability details for
Quantity
  • Availability
  • Add to Cart