C12H316CF3C6 Chemcia Scientific, LLC

N-(4-Amino-2-bromo-5-fluoro-phenyl)-2,2,2-trifluoro-acetamide

  • MDL number MFCD30723607
  • Molecular Weight : 301.03945

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8646

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8646
Availability details for
Quantity
  • Availability
  • Add to Cart