C12H316CF3D4 Chemcia Scientific, LLC

4-Bromo-5-methyl-2-nitro-benzoic acid

  • MDL number MFCD28127184
  • Cas Number : 1708958-91-1
  • Molecular Weight : 260.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-5-methyl-2-nitrobenzoic acid
Inchi Code 1S/C8H6BrNO4/c1-4-2-5(8(11)12)7(10(13)14)3-6(4)9/h2-3H,1H3,(H,11,12)
Inchi Key WKLFXBXUSBBVQO-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Pink Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8685

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8685
Availability details for
Quantity
  • Availability
  • Add to Cart