C12H316CF3E9 Chemcia Scientific, LLC

(2-Methoxy-6-methyl-pyridin-3-yl)-methanol

  • MDL number MFCD16611531
  • Cas Number : 351410-45-2
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methoxy-6-methyl-3-pyridinyl)methanol
Inchi Code 1S/C8H11NO2/c1-6-3-4-7(5-10)8(9-6)11-2/h3-4,10H,5H2,1-2H3
Inchi Key OWDFCQPJTQLELB-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8769

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8769
Availability details for
Quantity
  • Availability
  • Add to Cart