C12H316CF3F1 Chemcia Scientific, LLC

[1,2,4]Triazolo[1,5-a]pyridine-8-carboxylic acid methyl ester

  • MDL number MFCD11975619
  • Cas Number : 1259224-04-8
  • Molecular Weight : 177.16389

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8838

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8838
Availability details for
Quantity
  • Availability
  • Add to Cart