C12H316CF3F2 Chemcia Scientific, LLC

2-Amino-[1,2,4]triazolo[1,5-a]pyridine-5-carboxylic acid methyl ester

  • MDL number MFCD11975670
  • Cas Number : 1214902-53-0
  • Molecular Weight : 192.17856

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8843

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8843
Availability details for
Quantity
  • Availability
  • Add to Cart