C12H316CF3F3 Chemcia Scientific, LLC

2-Bromo-[1,2,4]triazolo[1,5-a]pyridine-5-carboxylic acid methyl ester

  • MDL number MFCD27952306
  • Cas Number : 1214902-64-3
  • Molecular Weight : 256.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-bromo-[1,2,4]triazolo[1,5-a]pyridine-5-carboxylate
Inchi Code 1S/C8H6BrN3O2/c1-14-7(13)5-3-2-4-6-10-8(9)11-12(5)6/h2-4H,1H3
Inchi Key YBTXISNQOPMXFF-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8844

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8844
Availability details for
Quantity
  • Availability
  • Add to Cart