C12H316CF3FE Chemcia Scientific, LLC

(4-Methyl-5-nitro-pyridin-2-yl)-hydrazine

  • MDL number MFCD09999892
  • Cas Number : 15367-24-5
  • Molecular Weight : 168.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-hydrazino-4-methyl-5-nitropyridine
Inchi Code 1S/C6H8N4O2/c1-4-2-6(9-7)8-3-5(4)10(11)12/h2-3H,7H2,1H3,(H,8,9)
Inchi Key LCBASHANKPYGSP-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Green Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8853

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8853
Availability details for
Quantity
  • Availability
  • Add to Cart