C12H316CF404 Chemcia Scientific, LLC

4-Bromo-6-methoxy-1,3-dihydro-benzoimidazol-2-one

  • MDL number MFCD22562190
  • Cas Number : 1388064-25-2
  • Molecular Weight : 243.06119

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8915

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8915
Availability details for
Quantity
  • Availability
  • Add to Cart