C12H316CF405 Chemcia Scientific, LLC

6-Methoxy-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-1,3-dihydro-benzoimidazol-2-one

  • MDL number MFCD30723662
  • Molecular Weight : 290.12953

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8916

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8916
Availability details for
Quantity
  • Availability
  • Add to Cart