C12H316CF407 Chemcia Scientific, LLC

7-Bromo-3H-benzoimidazole-5-carboxylic acid methyl ester

  • MDL number MFCD22421935
  • Cas Number : 1354756-19-6
  • Molecular Weight : 255.07234

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8920

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8920
Availability details for
Quantity
  • Availability
  • Add to Cart