C12H316CF409 Chemcia Scientific, LLC

(6-Amino-3-bromo-2-fluoro-phenyl)-methanol

  • MDL number MFCD30723666
  • Cas Number : 1036756-04-3
  • Molecular Weight : 220.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-amino-3-bromo-2-fluorophenyl)methanol
Inchi Code 1S/C7H7BrFNO/c8-5-1-2-6(10)4(3-11)7(5)9/h1-2,11H,3,10H2
Inchi Key FTGNJLGMELWIIK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8931

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8931
Availability details for
Quantity
  • Availability
  • Add to Cart