C12H316CF40A Chemcia Scientific, LLC

6-Amino-3-bromo-2-fluoro-benzoic acid methyl ester

  • MDL number MFCD30723667
  • Cas Number : 1637774-20-9
  • Molecular Weight : 248.05289

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8932

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8932
Availability details for
Quantity
  • Availability
  • Add to Cart