C12H316CF40C Chemcia Scientific, LLC

5-Bromo-3,4-dichloro-pyridin-2-ylamine

  • MDL number MFCD29761317
  • Cas Number : 1607837-98-8
  • Molecular Weight : 241.90306

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8940

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8940
Availability details for
Quantity
  • Availability
  • Add to Cart