C12H316CF414 Chemcia Scientific, LLC

3-Chloro-2-methoxy-benzonitrile

  • MDL number MFCD18393570
  • Cas Number : 725718-12-7
  • Molecular Weight : 167.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-2-methoxybenzonitrile
Inchi Code 1S/C8H6ClNO/c1-11-8-6(5-10)3-2-4-7(8)9/h2-4H,1H3
Inchi Key WWBDQIYXXXYFQJ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=8994

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8994
Availability details for
Quantity
  • Availability
  • Add to Cart