C12H316CF419 Chemcia Scientific, LLC

2,6-Dichloro-nicotinic acid tert-butyl ester

  • MDL number MFCD26396106
  • Cas Number : 1013647-75-0
  • Molecular Weight : 248.11067

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9066

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9066
Availability details for
Quantity
  • Availability
  • Add to Cart